Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MLKL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152649.3 | NP_689862.1 | 4 | 1320 | ||
XM_005255834.3 | XP_005255891.1 | 5 | 1219 | |||
XM_011522936.2 | XP_011521238.1 | 4 | 1359 | |||
GenBank mRNA | AK091708.1 | - | 4 | 1063 | ||
AL833362.1 | - | 3 | 1615 |