Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NLRC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178844.3 | NP_849172.2 | 8 | 2540 | ||
NR_075083.2 | 8 | 2601 | ||||
XM_017023027.1 | XP_016878516.1 | 8 | 2570 | |||
XM_017023028.1 | XP_016878517.1 | 9 | 2744 | |||
XM_017023029.1 | XP_016878518.1 | 10 | 2825 | |||
XM_017023030.1 | XP_016878519.1 | 10 | 2886 | |||
XM_017023031.1 | XP_016878520.1 | 8 | 2631 | |||
XM_017023032.1 | XP_016878521.1 | 10 | 3587 | |||
XM_017023033.1 | XP_016878522.1 | 10 | 2764 | |||
XM_017023034.1 | XP_016878523.1 | 9 | 2683 | |||
XM_017023035.1 | XP_016878524.1 | 9 | 2712 | |||
XM_017023036.1 | XP_016878525.1 | 8 | 3271 | |||
XM_017023037.1 | XP_016878526.1 | 9 | 2465 | |||
XM_017023038.1 | XP_016878527.1 | 9 | 2438 | |||
XM_017023039.1 | XP_016878528.1 | 8 | 2570 | |||
GenBank mRNA | AF501291.1 | - | 1 | 17 | ||
AF501292.1 | - | 1 | 17 | |||
AK074109.1 | - | 4 | 418 | |||
AK090431.1 | - | 8 | 2597 | |||
AK090476.1 | - | 8 | 3655 | |||
AY601811.1 | - | 8 | 2527 | |||
FJ889357.1 | - | 5 | 2116 |