Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC21A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105513.2 | NP_001098983.2 | 16 | 2338 | ||
NM_145755.2 | NP_665698.2 | 17 | 2482 | |||
XM_005264921.4 | XP_005264978.1 | 17 | 2411 | |||
XM_005264922.4 | XP_005264979.1 | 17 | 2411 | |||
XM_005264923.3 | XP_005264980.1 | 17 | 2411 | |||
XM_005264925.4 | XP_005264982.1 | 16 | 2288 | |||
XM_005264927.4 | XP_005264984.1 | 13 | 1865 | |||
XM_006713011.3 | XP_006713074.1 | 17 | 2411 | |||
XM_011533447.2 | XP_011531749.1 | 16 | 2312 | |||
GenBank mRNA | AJ487015.1 | - | 17 | 2444 | ||
AK093313.1 | - | 7 | 1022 | |||
AK302408.1 | - | 16 | 2321 | |||
AK302465.1 | - | 16 | 2192 | |||
BC129948.1 | - | 17 | 2393 | 40126472 |