Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: U2AF1L4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040425.2 | NP_001035515.1 | 2 | 108 | ||
NM_144987.3 | NP_659424.2 | 2 | 108 | |||
NR_110173.1 | 2 | 108 | ||||
GenBank mRNA | AK094213.1 | - | 2 | 80 | ||
AY569437.3 | - | 2 | 77 | |||
BC021186.1 | - | 2 | 77 | 5260387 | ||
BM696851.1 | - | 2 | 101 | |||
CD175846.1 | - | 2 | 117 |
Gene Symbol: LINC01412 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | HY038051.1 | - | 443 |