Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STOM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270526.1 | NP_001257455.1 | 7 | 785 | ||
NM_004099.5 | NP_004090.4 | 7 | 876 | |||
NM_198194.2 | NP_937837.1 | 3 | 381 | |||
NR_073037.1 | 8 | 1006 | ||||
GenBank mRNA | AK302977.1 | - | 7 | 802 | ||
AK304449.1 | - | 8 | 950 | |||
AK304518.1 | - | 6 | 517 | |||
AK307999.1 | - | 7 | 819 | |||
BC010703.1 | - | 7 | 798 | 3873494 | ||
BG701899.1 | - | 3 | 334 | |||
CR542100.1 | - | 7 | 759 | |||
M81635.1 | - | 7 | 780 | |||
X60067.1 | - | 7 | 759 |