Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXLNA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175852.3 | NP_787048.1 | 9, 10 | 1323 | ||
XM_011540931.2 | XP_011539233.1 | 8, 9 | 1246 | |||
XM_011540932.2 | XP_011539234.1 | 7, 8 | 1083 | |||
XM_017000561.1 | XP_016856050.1 | 8, 9 | 1715 | |||
XM_017000562.1 | XP_016856051.1 | 8, 9 | 1616 | |||
XM_017000563.1 | XP_016856052.1 | 9, 10 | 1497 | |||
XM_017000564.1 | XP_016856053.1 | 8, 9 | 1177 | |||
XM_017000565.1 | XP_016856054.1 | 8, 9 | 1271 | |||
GenBank mRNA | AF516206.1 | - | 8, 9 | 1336 | ||
AL832338.1 | - | 9, 10 | 1360 | |||
AL832636.2 | - | 9, 10 | 1382 | |||
AL832637.2 | - | 8, 9 | 1539 | |||
BC046565.1 | - | 1, 2 | 35 | |||
BC080578.1 | - | 8, 9 | 1300 | 6598234 | ||
BC103823.1 | - | 8, 9 | 1288 | 40001224 | ||
BC103824.1 | - | 8, 9 | 1288 | 40001225 |