Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPAG17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_206996.2 | NP_996879.1 | 30 | 4404 | ||
XM_006710426.3 | XP_006710489.1 | 29 | 4290 | |||
XM_006710427.3 | XP_006710490.1 | 29 | 4209 | |||
XM_006710428.3 | XP_006710491.1 | 17 | 2475 | |||
XM_006710429.3 | XP_006710492.1 | 14 | 2087 | |||
XM_006710431.3 | XP_006710494.1 | 11 | 1534 | |||
XM_011540934.1 | XP_011539236.1 | 30 | 4404 | |||
XM_011540935.2 | XP_011539237.1 | 29 | 4243 | |||
XM_011540936.2 | XP_011539238.1 | 21 | 3062 | |||
XM_011540937.2 | XP_011539239.1 | 21 | 3228 | |||
XM_011540939.2 | XP_011539241.1 | 18 | 2513 | |||
XM_011540941.2 | XP_011539243.1 | 17 | 2523 | |||
XM_011540942.2 | XP_011539244.1 | 17 | 2331 | |||
GenBank mRNA | AK126319.1 | - | 7 | 1002 | ||
AL833485.1 | - | 16 | 2174 | |||
AY555274.1 | - | 30 | 4390 | |||
BX648110.1 | - | 30 | 4403 |