Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPRED2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128210.1 | NP_001121682.1 | 3 | 466 | ||
NM_181784.2 | NP_861449.2 | 3 | 819 | |||
XM_005264200.4 | XP_005264257.2 | 3 | 896 | |||
XM_005264202.4 | XP_005264259.1 | 3 | 896 | |||
GenBank mRNA | AK056479.1 | - | 3 | 383 | ||
AK299079.1 | - | 3 | 319 | |||
AY299090.1 | - | 3 | 480 | |||
BC029300.1 | - | 3 | 396 | |||
BC111495.1 | - | 3 | 807 | |||
BC130292.1 | - | 3 | 416 | 40146323 | ||
BC136334.1 | - | 3 | 416 | 9020321 | ||
DC335282.1 | - | 3 | 479 |