Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C3orf67 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198463.2 | NP_940865.1 | 6 | 512 | ||
XM_005264929.2 | XP_005264986.1 | 6 | 892 | |||
XM_006713013.3 | XP_006713076.1 | 6 | 892 | |||
XM_011533451.2 | XP_011531753.1 | 6 | 892 | |||
XM_011533453.2 | XP_011531755.1 | 6 | 892 | |||
XM_011533455.2 | XP_011531757.1 | 6 | 892 | |||
XM_011533456.1 | XP_011531758.1 | 6 | 601 | |||
XM_011533457.1 | XP_011531759.1 | 4 | 381 | |||
XM_011533458.2 | XP_011531760.1 | 3 | 264 | |||
XM_011533459.2 | XP_011531761.1 | 6 | 892 | |||
XM_011533460.2 | XP_011531762.1 | 1 | 21 | |||
GenBank mRNA | AK124111.1 | - | 6 | 514 | ||
AK296111.1 | - | 6 | 606 | |||
BC050317.1 | - | 6 | 879 | |||
BC132815.1 | - | 4 | 326 | 40146837 | ||
BC140002.1 | - | 7 | 622 | |||
BC151142.1 | - | 4 | 326 | 9020498 |