Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174919.3 | NP_777579.2 | 4 | 1305 | ||
GenBank mRNA | AF258593.1 | - | 5 | 2247 | ||
AK290650.1 | - | 4 | 1256 | |||
BC067345.1 | - | 4 | 1305 | 6084901 |