Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAMD14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257359.1 | NP_001244288.1 | 5 | 928 | ||
NM_174920.3 | NP_777580.1 | 5 | 928 | |||
XM_011524490.2 | XP_011522792.1 | 2 | 114 | |||
XM_017024322.1 | XP_016879811.1 | 4 | 1312 | |||
XM_017024323.1 | XP_016879812.1 | 6 | 1095 | |||
XM_017024324.1 | XP_016879813.1 | 5 | 1783 | |||
GenBank mRNA | AK094209.1 | - | 5 | 859 | ||
AK095809.1 | - | 2 | 229 | |||
AK225767.1 | - | 5 | 859 | |||
AK291482.1 | - | 5 | 855 | |||
AY148482.1 | - | 5 | 849 | |||
BC029872.1 | - | 1 | 40 | |||
BC047081.1 | - | 4 | 582 | |||
BC136485.1 | - | 5 | 841 | 9020474 | ||
BC141822.1 | - | 5 | 868 | 40148162 | ||
BC142728.1 | - | 5 | 825 | 40148957 |