Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAPT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153365.2 | NP_699196.2 | 9 | 1107 | ||
XM_005248139.2 | XP_005248196.1 | 8 | 921 | |||
XM_005248140.3 | XP_005248197.1 | 8 | 871 | |||
XM_011513812.2 | XP_011512114.1 | 8 | 970 | |||
XM_011513815.2 | XP_011512117.1 | 8 | 2206 | |||
XM_011513816.2 | XP_011512118.1 | 8 | 915 | |||
XM_011513817.2 | XP_011512119.1 | 6 | 484 | |||
XM_017007876.1 | XP_016863365.1 | 10 | 997 | |||
GenBank mRNA | AK074494.1 | - | 9 | 1040 | ||
AK296104.1 | - | 8 | 909 | |||
AK299163.1 | - | 5 | 403 | |||
BC066899.1 | - | 9 | 1045 | 4825556 |