Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CMYA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153610.4 | NP_705838.3 | 2 | 5483 | ||
XM_017009212.1 | XP_016864701.1 | 2 | 5351 | |||
GenBank mRNA | AF177292.1 | - | 1 | 206 | ||
AK096453.1 | - | 1 | 1570 | |||
BC022422.1 | - | 1 | 602 | |||
BX647452.1 | - | 1 | 1629 |