Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM219A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184940.1 | NP_001171869.1 | 4 | 638 | ||
NM_001184941.1 | NP_001171870.1 | 4 | 638 | |||
NM_001184942.1 | NP_001171871.1 | 4 | 605 | |||
NM_001184943.1 | NP_001171872.1 | 4 | 587 | |||
NM_001184945.1 | NP_001171874.1 | 4 | 554 | |||
NM_147202.1 | NP_671735.1 | 4 | 587 | |||
XM_005251402.4 | XP_005251459.1 | 5 | 845 | |||
XM_011517797.2 | XP_011516099.1 | 4 | 645 | |||
XM_011517798.2 | XP_011516100.1 | 4 | 594 | |||
XM_011517799.2 | XP_011516101.1 | 4 | 443 | |||
GenBank mRNA | AK096350.1 | - | 4 | 587 | ||
AK123887.1 | - | 5 | 449 | |||
AK294054.1 | - | 4 | 605 | |||
AK299882.1 | - | 4 | 633 | |||
BC041009.2 | - | 4 | 554 | 5284905 | ||
DA110626.1 | - | 4 | 377 |