Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CT83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017978.3 | NP_001017978.1 | 1, 2 | 245 | ||
GenBank mRNA | AK000618.1 | - | 1, 2 | 215 | ||
AK026566.1 | - | 1, 2 | 215 | |||
BC062223.1 | - | 1, 2 | 192 | 6101827 | ||
HY170569.1 | - | 1, 2 | 246 |