Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CERS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290341.2 | NP_001277270.1 | 13 | 1559 | ||
NM_001290342.2 | NP_001277271.1 | 12 | 1385 | |||
NM_001290343.1 | NP_001277272.1 | 12 | 1296 | |||
NM_178842.4 | NP_849164.2 | 12 | 1292 | |||
XM_011521355.2 | XP_011519657.1 | 13 | 1602 | |||
XM_011521357.2 | XP_011519659.1 | 13 | 1496 | |||
XM_011521358.1 | XP_011519660.1 | 13 | 1380 | |||
XM_017022002.1 | XP_016877491.1 | 12 | 1376 | |||
XM_017022003.1 | XP_016877492.1 | 12 | 1301 | |||
XM_017022004.1 | XP_016877493.1 | 11 | 1212 | |||
GenBank mRNA | BC028703.1 | - | 12 | 1373 | 4837657 | |
BC034500.1 | - | 12 | 1248 | 4838423 | ||
BC034970.1 | - | 12 | 1294 | 4822006 |