Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NLRP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297743.1 | NP_001284672.1 | 2 | 1575 | ||
NM_145007.3 | NP_659444.2 | 5 | 2425 | |||
GenBank mRNA | AK090621.1 | - | 1 | 625 | ||
AK129784.1 | - | 1 | 652 | |||
AK310394.1 | - | 5 | 2102 | |||
AY095145.1 | - | 3 | 1813 | |||
AY154466.1 | - | 2 | 1714 | |||
AY168967.1 | - | 3 | 1802 | |||
AY168968.1 | - | 2 | 1474 | |||
AY226382.1 | - | 2 | 1714 | |||
BC034730.1 | - | 3 | 1802 | 4341197 |