Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C3orf58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134470.1 | NP_001127942.1 | 3 | 565 | ||
NM_173552.3 | NP_775823.1 | 3 | 1519 | |||
XM_011512543.2 | XP_011510845.1 | 2 | 1104 | |||
XM_017005869.1 | XP_016861358.1 | 3 | 399 | |||
GenBank mRNA | AK095161.1 | - | 3 | 581 | ||
AK095400.1 | - | 3 | 425 | |||
AK294020.1 | - | 3 | 463 | |||
AK315082.1 | - | 3 | 1493 | |||
AL832839.1 | - | 3 | 581 | |||
BC037293.2 | - | 3 | 1246 | 5267770 | ||
DC310421.1 | - | 3 | 463 |