Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPOR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000121.3 | NP_000112.1 | 8 | 1718 | ||
NR_033663.1 | 8 | 1968 | ||||
GenBank mRNA | BC112153.1 | - | 8 | 1660 | 8327621 | |
BC143697.1 | - | 9 | 1796 | |||
M34986.1 | - | 8 | 1583 | |||
M60459.1 | - | 8 | 1688 |