Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MED19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317078.1 | NP_001304007.1 | 3 | 674 | ||
NM_153450.2 | NP_703151.2 | 3 | 674 | |||
GenBank mRNA | AY148462.1 | - | 3 | 403 | ||
BC009723.1 | - | 3 | 550 | |||
BC037223.1 | - | 3 | 552 | 5197938 | ||
EB386171.1 | - | 3 | 560 | |||
U92995.1 | - | 2 | 108 |