Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBATA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318241.1 | NP_001305170.1 | 10, 11 | 1379 | ||
NM_001318242.1 | NP_001305171.1 | 10, 11 | 1443 | |||
NM_001318243.1 | NP_001305172.1 | 10, 11 | 1373 | |||
NM_152710.3 | NP_689923.3 | 10, 11 | 1376 | |||
NR_134531.1 | 9, 10 | 1294 | ||||
NR_134532.1 | 10, 11 | 1302 | ||||
NR_134533.1 | 6, 7 | 912 | ||||
NR_134534.1 | 5, 6 | 836 | ||||
XM_017015848.1 | XP_016871337.1 | 11, 12 | 1503 | |||
XM_017015849.1 | XP_016871338.1 | 11, 12 | 1549 | |||
XM_017015853.1 | XP_016871342.1 | 11, 12 | 1454 | |||
XM_017015860.1 | XP_016871349.1 | 9, 10 | 1227 | |||
GenBank mRNA | AK057382.1 | - | 10, 11 | 1358 | ||
BC137543.1 | - | 10, 11 | 1410 | 9021543 | ||
BC137544.1 | - | 10, 11 | 1344 | 9021544 | ||
BC139728.1 | - | 9, 10 | 1243 | 40036949 | ||
BC144692.1 | - | 10, 11 | 1341 | 9053226 | ||
BC144694.1 | - | 10, 11 | 1347 | 9053228 |