Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPS15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159969.1 | NP_001153441.1 | 10, 11 | 1417 | ||
NM_001981.2 | NP_001972.1 | 22, 23 | 2286 | |||
XM_005270618.2 | XP_005270675.1 | 21, 22 | 2227 | |||
XM_017000615.1 | XP_016856104.1 | 22, 23 | 2428 | |||
XM_017000616.1 | XP_016856105.1 | 21, 22 | 2224 | |||
XM_017000617.1 | XP_016856106.1 | 21, 22 | 2113 | |||
XM_017000618.1 | XP_016856107.1 | 20, 21 | 2023 | |||
XM_017000619.1 | XP_016856108.1 | 23, 24 | 2310 | |||
XM_017000620.1 | XP_016856109.1 | 22, 23 | 2477 | |||
GenBank mRNA | AK090936.1 | - | 10, 11 | 1143 | ||
AK294297.1 | - | 11, 12 | 1266 | |||
AK308128.1 | - | 6, 7 | 703 | |||
AK313396.1 | - | 22, 23 | 2235 | |||
BC054006.1 | - | 21, 22 | 1802 | 6164633 | ||
BX647676.1 | - | 10, 11 | 1398 | |||
DQ367924.1 | - | 22, 23 | 2194 | |||
U07707.1 | - | 22, 23 | 2215 | |||
Z29064.1 | - | 22, 23 | 2286 |