Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOK6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152721.5 | NP_689934.2 | 8 | 1067 | ||
XM_011525875.2 | XP_011524177.1 | 8 | 1103 | |||
XM_017025610.1 | XP_016881099.1 | 6 | 3128 | |||
XM_017025611.1 | XP_016881100.1 | 6 | 1642 | |||
GenBank mRNA | AK057795.1 | - | 7 | 776 | ||
AY599248.1 | - | 8 | 1065 | |||
BC008583.1 | - | 6 | 649 | |||
BC019045.2 | - | 6 | 692 | |||
BC096744.1 | - | 8 | 1060 | 5260167 |