Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNRNPA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194247.2 | NP_919223.1 | 3 | 408 | ||
XM_005246380.1 | XP_005246437.1 | 3 | 408 | |||
XM_005246381.4 | XP_005246438.1 | 3 | 314 | |||
XM_006712365.2 | XP_006712428.1 | 3 | 408 | |||
XM_017003587.1 | XP_016859076.1 | 3 | 314 | |||
XM_017003588.1 | XP_016859077.1 | 3 | 408 | |||
GenBank mRNA | AK293125.1 | - | 3 | 276 | ||
AK304687.1 | - | 3 | 271 | |||
AK316434.1 | - | 3 | 276 | |||
AY363225.1 | - | 3 | 302 | |||
BC064494.1 | - | 3 | 320 | |||
BC112027.1 | - | 3 | 330 | 8327495 | ||
BC113470.1 | - | 3 | 330 | 8322522 | ||
BX647217.1 | - | 3 | 265 | |||
DB093606.1 | - | 3 | 408 |