Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARL5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178815.3 | NP_848930.1 | 4 | 555 | ||
XM_005252401.4 | XP_005252458.1 | 4 | 420 | |||
GenBank mRNA | AF494061.1 | - | 4 | 477 | ||
AK027275.1 | - | 4 | 508 | |||
BC024163.1 | - | 4 | 535 | 3931095 |