Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRN4CL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203422.3 | NP_981967.1 | 2 | 1406 | ||
GenBank mRNA | AK291334.1 | - | 2 | 1037 | ||
AL834319.1 | - | 2 | 1235 | |||
AY358354.1 | - | 2 | 1079 | |||
BC053902.1 | - | 2 | 1109 | 6165542 |