Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRSS54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080492.1 | NP_001073961.1 | 6 | 960 | ||
NM_001305173.1 | NP_001292102.1 | 6 | 957 | |||
NM_001305174.1 | NP_001292103.1 | 5 | 782 | |||
GenBank mRNA | AK058068.1 | - | 6 | 902 | ||
AK302175.1 | - | 5 | 712 | |||
AL137484.1 | - | 2 | 250 | |||
BC057843.1 | - | 6 | 949 | 5296090 |