Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OPN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181744.3 | NP_859528.1 | 5 | 928 | ||
NR_033806.1 | 5 | 945 | ||||
XM_017010409.1 | XP_016865898.1 | 3 | 1799 | |||
XM_017010410.1 | XP_016865899.1 | 5 | 895 | |||
XM_017010411.1 | XP_016865900.1 | 4 | 1036 | |||
XM_017010412.1 | XP_016865901.1 | 4 | 1092 | |||
XM_017010413.1 | XP_016865902.1 | 4 | 749 | |||
XM_017010414.1 | XP_016865903.1 | 4 | 1645 | |||
XM_017010415.1 | XP_016865904.1 | 5 | 945 | |||
XM_017010416.1 | XP_016865905.1 | 5 | 930 | |||
GenBank mRNA | AY288419.1 | - | 5 | 891 | ||
AY377391.1 | - | 5 | 905 | |||
BC126194.1 | - | 5 | 928 | 8991910 | ||
BC126198.1 | - | 5 | 928 | 8991914 | ||
BX647224.1 | - | 5 | 887 |
Set Membership: |
G-protein coupled receptor |