Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289020.1 | NP_001275949.1 | 8 | 977 | ||
NM_001289021.1 | NP_001275950.1 | 8 | 977 | |||
NM_145027.4 | NP_659464.3 | 8 | 977 | |||
XM_005248904.4 | XP_005248961.1 | 8 | 977 | |||
XM_011514357.2 | XP_011512659.1 | 8 | 902 | |||
XM_011514358.2 | XP_011512660.1 | 8 | 902 | |||
XM_011514359.2 | XP_011512661.1 | 8 | 902 | |||
XM_011514361.2 | XP_011512663.1 | 8 | 977 | |||
XM_017010427.1 | XP_016865916.1 | 7 | 867 | |||
XM_017010428.1 | XP_016865917.1 | 3 | 327 | |||
GenBank mRNA | AK131471.1 | - | 8 | 977 | ||
BC117307.1 | - | 8 | 907 | 40125858 | ||
BC143444.1 | - | 8 | 907 | 9051952 | ||
BC143445.1 | - | 8 | 907 | 9051953 | ||
BX649045.1 | - | 5 | 557 |