Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LEMD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143944.1 | NP_001137416.1 | 2, 3 | 289 | ||
NM_181336.3 | NP_851853.1 | 3, 4 | 870 | |||
GenBank mRNA | AK074959.1 | - | 3, 4 | 923 | ||
AK301083.1 | - | 2, 3 | 283 | |||
BC051803.1 | - | 3, 4 | 860 | 5240212 | ||
DC340017.1 | - | 2, 3 | 348 |