Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LEMD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143944.1 | NP_001137416.1 | 4 | 407 | ||
NM_181336.3 | NP_851853.1 | 5 | 988 | |||
GenBank mRNA | AK074959.1 | - | 5 | 1041 | ||
AK127084.1 | - | 2 | 1217 | |||
AK301083.1 | - | 4 | 401 | |||
AK309888.1 | - | 1 | 165 | |||
BC051803.1 | - | 5 | 978 | 5240212 | ||
DC340017.1 | - | 4 | 466 |