Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079653.1 | NP_001073121.1 | 13 | 1901 | ||
NM_152744.3 | NP_689957.3 | 39 | 5756 | |||
XM_011515188.1 | XP_011513490.1 | 33 | 4597 | |||
XM_011515189.1 | XP_011513491.1 | 33 | 4594 | |||
XM_017011837.1 | XP_016867326.1 | 33 | 4597 | |||
GenBank mRNA | AK074077.1 | - | 6 | 696 | ||
AK074083.1 | - | 24 | 3392 | |||
AL833354.1 | - | 13 | 1898 | |||
AY310398.1 | - | 39 | 5756 |