Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004450.2 | NP_004441.1 | 4 | 336 | ||
GenBank mRNA | AK130850.1 | - | 4 | 322 | ||
AK312186.1 | - | 4 | 331 | |||
BC014301.2 | - | 4 | 319 | 3507241 | ||
BC071709.1 | - | 4 | 283 | 5503738 | ||
BT006877.1 | - | 3 | 265 | |||
CR450298.1 | - | 3 | 265 | |||
CR542178.1 | - | 3 | 265 | |||
D85758.1 | - | 4 | 339 | |||
U66871.1 | - | 4 | 336 |