Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRWD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317721.1 | NP_001304650.1 | 2 | 375 | ||
NM_152892.2 | NP_690852.1 | 2 | 357 | |||
XM_017011845.1 | XP_016867334.1 | 2 | 258 | |||
GenBank mRNA | AB209379.1 | - | 2 | 235 | ||
AK290967.1 | - | 2 | 256 | |||
AK313771.1 | - | 2 | 240 | |||
AL133057.1 | - | 2 | 307 | |||
BC009436.1 | - | 1 | 48 | |||
BC030547.1 | - | 2 | 297 | 5170588 | ||
HY037356.1 | - | 2 | 357 | |||
HY051509.1 | - | 2 | 344 |