Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERV3-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007253.3 | NP_001007254.2 | 2 | 940 | ||
GenBank mRNA | AK295189.1 | - | 2 | 847 |