Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPRC6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286354.1 | NP_001273283.1 | 6 | 1911 | ||
NM_001286355.1 | NP_001273284.1 | 5 | 2223 | |||
NM_148963.3 | NP_683766.2 | 6 | 2436 | |||
XM_017010475.1 | XP_016865964.1 | 7 | 4369 | |||
GenBank mRNA | AF502962.1 | - | 6 | 2414 | ||
AY435125.1 | - | 6 | 2425 | |||
AY435126.1 | - | 6 | 1900 | |||
AY435127.1 | - | 5 | 2212 |
Set Membership: |
G-protein coupled receptor |