Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NKAPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007531.2 | NP_001007532.1 | 1 | 679 | ||
GenBank mRNA | AF085870.1 | - | 1 | 432 | ||
BC038240.1 | - | 1 | 609 | 5766894 | ||
BC101681.1 | - | 1 | 637 | 8069187 | ||
BC101685.1 | - | 1 | 637 | 8069191 |