Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DEFB110 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037728.2 | NP_001032817.1 | 2 | 116 | ||
GenBank mRNA | DQ012014.1 | - | 2 | 70 |