Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199779.1 | NP_001186708.1 | 2, 3 | 266 | ||
NM_001199780.1 | NP_001186709.1 | 1, 2 | 110 | |||
NM_002794.4 | NP_002785.1 | 2, 3 | 634 | |||
GenBank mRNA | AB209470.1 | - | 2, 3 | 320 | ||
AK296956.1 | - | 2, 3 | 241 | |||
AK310023.1 | - | 1, 2 | 110 | |||
AV762678.2 | - | 2, 3 | 328 | |||
BC000268.1 | - | 2, 3 | 280 | |||
BC101836.1 | - | 2, 3 | 266 | 8069342 | ||
BC105126.1 | - | 2, 3 | 266 | 8144129 | ||
BC107901.1 | - | 2, 3 | 281 | 6745302 | ||
BM849239.1 | - | 2, 3 | 266 | |||
BT007137.1 | - | 2, 3 | 222 | |||
CR456862.1 | - | 2, 3 | 222 | |||
D26599.1 | - | 2, 3 | 251 |