Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PELI3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098510.1 | NP_001091980.1 | 4 | 511 | ||
NM_001243135.1 | NP_001230064.1 | 4 | 350 | |||
NM_001243136.1 | NP_001230065.1 | 3 | 358 | |||
NM_145065.2 | NP_659502.2 | 5 | 583 | |||
XM_011544884.2 | XP_011543186.1 | 5 | 586 | |||
XM_017017465.1 | XP_016872954.1 | 3 | 335 | |||
GenBank mRNA | AF487456.1 | - | 4 | 419 | ||
AF487457.1 | - | 3 | 347 | |||
AK074201.1 | - | 5 | 521 | |||
AK075253.1 | - | 1 | 23 | |||
AK094060.1 | - | 4 | 350 | |||
AL834395.1 | - | 5 | 530 | |||
BC021256.2 | - | 5 | 524 | 3535985 | ||
BC025723.1 | - | 4 | 432 | 5198999 | ||
BX341733.2 | - | 3 | 295 |