Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FAM21C SIRNA Assays ›
Gene Symbol:
FAM21C
Gene Name:
family with sequence similarity 21 member C
Gene Aliases:
FAM21A, VPEF
Chromosome Location:
Chr.10: 45727170 - 45792964 on Build GRCh38
UniGene ID:
Hs.365286
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s48439
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FAM21C
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001169106.1
NP_001162577.1
29
4072
NM_001169107.1
NP_001162578.1
28
3970
NM_015262.2
NP_056077.2
30
4195
XM_011539566.1
XP_011537868.1
31
4225
XM_011539567.1
XP_011537869.1
30
4153
XM_011539568.1
XP_011537870.1
30
4138
XM_011539569.1
XP_011537871.1
30
4072
XM_011539570.2
XP_011537872.1
30
4114
XM_011539571.1
XP_011537873.1
28
3826
XM_011539573.2
XP_011537875.1
13
2351
XM_017016014.1
XP_016871503.1
30
4102
XM_017016015.1
XP_016871504.1
29
4090
XM_017016016.1
XP_016871505.1
29
4075
XM_017016017.1
XP_016871506.1
29
4066
XM_017016018.1
XP_016871507.1
29
4009
XM_017016019.1
XP_016871508.1
29
4000
XM_017016020.1
XP_016871509.1
28
3967
XM_017016021.1
XP_016871510.1
31
4153
XM_017016022.1
XP_016871511.1
30
4136
XM_017016023.1
XP_016871512.1
28
3885
XM_017016024.1
XP_016871513.1
19
2967
XM_017016025.1
XP_016871514.1
20
3038
GenBank mRNA
AK001545.1
-
13
2419
AK291430.1
-
30
4183
AK303048.1
-
28
3934
AL050279.1
-
4
1930
BC006456.1
-
4
1123
BC150611.1
-
29
4037
9056981
Back To Top
More Information
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
protein transport
retrograde transport, endosome to Golgi
regulation of substrate adhesion-dependent cell spreading
retrograde transport, endosome to plasma membrane
negative regulation of barbed-end actin filament capping
protein binding
phosphatidylinositol-4,5-bisphosphate binding
phosphatidylinositol-3,4,5-trisphosphate binding
phosphatidylinositol-5-phosphate binding
phosphatidylinositol-3-phosphate binding
phosphatidylinositol-3,4-bisphosphate binding
phosphatidylinositol-4-phosphate binding
phosphatidylinositol-3,5-bisphosphate binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0