Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASPHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181718.3 | NP_859069.2 | 3 | 1449 | ||
XM_017023107.1 | XP_016878596.1 | 4 | 1430 | |||
GenBank mRNA | BC009033.1 | - | 3 | 1176 | ||
BC029136.1 | - | 3 | 885 | |||
BC034275.3 | - | 3 | 922 | |||
BC040933.2 | - | 3 | 961 | |||
BC085011.1 | - | 3 | 1297 |