Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAALADL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207015.2 | NP_996898.2 | 2 | 443 | ||
XM_006713560.2 | XP_006713623.1 | 4 | 526 | |||
XM_011512612.2 | XP_011510914.1 | 2 | 3780 | |||
XM_011512613.1 | XP_011510915.1 | 2 | 588 | |||
XM_011512615.2 | XP_011510917.1 | 2 | 3781 | |||
XM_011512617.2 | XP_011510919.1 | 2 | 3778 | |||
XM_017006070.1 | XP_016861559.1 | 6 | 968 | |||
XM_017006071.1 | XP_016861560.1 | 7 | 1038 | |||
XM_017006072.1 | XP_016861561.1 | 8 | 1172 | |||
XM_017006073.1 | XP_016861562.1 | 6 | 771 | |||
XM_017006074.1 | XP_016861563.1 | 8 | 3190 | |||
XM_017006075.1 | XP_016861564.1 | 5 | 1915 | |||
XM_017006076.1 | XP_016861565.1 | 5 | 708 | |||
XM_017006077.1 | XP_016861566.1 | 7 | 2119 | |||
XM_017006078.1 | XP_016861567.1 | 5 | 1936 | |||
XM_017006079.1 | XP_016861568.1 | 3 | 1412 | |||
XM_017006080.1 | XP_016861569.1 | 3 | 1783 | |||
XM_017006081.1 | XP_016861570.1 | 2 | 3780 | |||
XM_017006082.1 | XP_016861571.1 | 5 | 1922 | |||
GenBank mRNA | AJ607396.1 | - | 2 | 484 | ||
AK308416.1 | - | 2 | 459 | |||
AL832144.1 | - | 2 | 443 | |||
BC057243.1 | - | 2 | 446 |