Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C4orf22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152770.2 | NP_689983.2 | 3, 4 | 392 | ||
XM_017007972.1 | XP_016863461.1 | 4, 5 | 544 | |||
XM_017007973.1 | XP_016863462.1 | 4, 5 | 547 | |||
XM_017007974.1 | XP_016863463.1 | 4, 5 | 552 | |||
XM_017007975.1 | XP_016863464.1 | 2, 3 | 532 | |||
XM_017007978.1 | XP_016863467.1 | 2, 3 | 409 | |||
GenBank mRNA | BC034296.1 | - | 3, 4 | 392 | 5165558 |