Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLA2G4F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_213600.3 | NP_998765.3 | 15 | 1713 | ||
NR_033151.1 | 14 | 1640 | ||||
GenBank mRNA | AK122623.1 | - | 15 | 1684 | ||
AK304368.1 | - | 15 | 1695 | |||
BC035335.1 | - | 14 | 1610 | |||
BC131531.1 | - | 15 | 1625 | 40036909 | ||
BC142682.1 | - | 15 | 1684 | 40148418 | ||
BC146648.1 | - | 15 | 1684 | 40147152 | ||
CR749451.1 | - | 8 | 3786 |