Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYADML2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145113.2 | NP_001138585.2 | 3 | 1075 | ||
GenBank mRNA | BC029306.1 | - | 3 | 1075 |