Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL6A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278298.1 | NP_001265227.1 | 38 | 7804 | ||
NM_153264.6 | NP_694996.5 | 38 | 7804 | |||
NR_022012.2 | 38 | 7804 | ||||
XM_011512621.2 | XP_011510923.1 | 37 | 7615 | |||
GenBank mRNA | AK093199.1 | - | 4 | 1025 | ||
AK123718.1 | - | 6 | 1615 | |||
AM906083.1 | - | 4 | 1139 | |||
AM906084.1 | - | 1 | 377 | |||
EU085556.1 | - | 38 | 7804 |