Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EVI2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003927.2 | NP_001003927.1 | 3 | 613 | ||
NM_014210.3 | NP_055025.2 | 2 | 472 | |||
GenBank mRNA | AK295311.1 | - | 3 | 548 | ||
AK312348.1 | - | 2 | 425 | |||
AV724051.1 | - | 2 | 393 | |||
BC035572.1 | - | 2 | 360 | 5229926 | ||
BG699778.1 | - | 3 | 513 | |||
BI597493.1 | - | 2 | 428 | |||
DA364537.1 | - | 2 | 472 |