Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039547.2 | NP_001034636.1 | 10 | 1076 | ||
NR_033289.1 | 11 | 2159 | ||||
XM_011512624.2 | XP_011510926.1 | 10 | 874 | |||
XM_017006088.1 | XP_016861577.1 | 10 | 1061 | |||
XM_017006089.1 | XP_016861578.1 | 11 | 1139 | |||
XM_017006090.1 | XP_016861579.1 | 9 | 987 | |||
GenBank mRNA | AK090901.1 | - | 1 | 125 | ||
AK127641.1 | - | 10 | 1044 | |||
BC032470.1 | - | 11 | 2118 | 5217372 | ||
BX648359.1 | - | 10 | 1056 | |||
BX648681.1 | - | 11 | 2096 |