Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EML3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300793.1 | NP_001287722.1 | 11, 12 | 1680 | ||
NM_001300794.1 | NP_001287723.1 | 11, 12 | 1677 | |||
NM_153265.2 | NP_694997.2 | 11, 12 | 1677 | |||
XM_005273878.3 | XP_005273935.1 | 11, 12 | 1935 | |||
XM_006718489.3 | XP_006718552.1 | 11, 12 | 2052 | |||
XM_006718490.3 | XP_006718553.1 | 11, 12 | 2052 | |||
XM_006718491.3 | XP_006718554.1 | 11, 12 | 2049 | |||
XM_011544896.2 | XP_011543198.1 | 11, 12 | 2049 | |||
XM_017017480.1 | XP_016872969.1 | 11, 12 | 1423 | |||
XM_017017481.1 | XP_016872970.1 | 10, 11 | 1309 | |||
XM_017017482.1 | XP_016872971.1 | 11, 12 | 1423 | |||
XM_017017483.1 | XP_016872972.1 | 11, 12 | 1480 | |||
GenBank mRNA | AK093146.1 | - | 10, 11 | 1352 | ||
AK128679.1 | - | 10, 11 | 2094 | |||
AK296901.1 | - | 11, 12 | 1442 | |||
AK310579.1 | - | 13, 14 | 1503 | |||
AL833388.1 | - | 8, 9 | 932 | |||
BC075824.1 | - | 10, 11 | 1283 | |||
BC108280.1 | - | 11, 12 | 1461 | 3915493 |